missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD2 Mouse anti-Human, Porcine, Rat, Clone: 2G1G10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66302-1-IG-20UL
This item is not returnable.
View return policy
Description
Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).Specifications
| CHCHD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y6H1 | |
| Chchd2 | |
| CHCHD2 Fusion Protein Ag24219 | |
| 20 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2G1G10 | |
| Unconjugated | |
| CHCHD2 | |
| C7orf17, CHCHD2, NS2TP | |
| Mouse | |
| Protein G | |
| RUO | |
| 100518545, 316643, 51142 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction