missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAF1B Antibody (SS 53), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
£161.00 - £362.00
Specifications
| Antigen | CHAF1B |
|---|---|
| Clone | SS 53 |
| Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, KnockDown Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109039
|
Novus Biologicals
NB500-212SS |
0.025 mL |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169789
|
Novus Biologicals
NB500-212 |
0.1 mL |
£362.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHAF1B Monoclonal specifically detects CHAF1B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.Specifications
| CHAF1B | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, KnockDown Validated | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Q13112 | |
| 8208 | |
| Recombinant human CHAF1B [UniProt# Q13112] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| SS 53 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research | |
| Ascites with 0.1% Sodium Azide | |
| CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7 | |
| CHAF1B | |
| IgG1 | |
| Unpurified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title