missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAF1B Antibody (SS 53), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB500-212SS
This item is not returnable.
View return policy
Description
CHAF1B Monoclonal specifically detects CHAF1B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.
Specifications
| CHAF1B | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7 | |
| Mouse | |
| 60 kDa | |
| 0.025 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research | |
| 8208 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| SS 53 | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, KnockDown Validated | |
| Q13112 | |
| CHAF1B | |
| Recombinant human CHAF1B [UniProt# Q13112] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction