missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAF1A Rabbit anti-Human, Clone: 1T1O5, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | CHAF1A |
|---|---|
| Clone | 1T1O5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18322902
|
Bio-Techne
NBP3-16690-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18356215
|
Novus Biologicals
NBP3-16690-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHAF1A Monoclonal antibody specifically detects CHAF1A in Human samples. It is validated for Western BlotSpecifications
| CHAF1A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10036 | |
| IgG | |
| Affinity purified |
| 1T1O5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAF, CAF1, CAF-1, CAF-1 subunit A, CAF1B, CAF1P150CAF-I 150 kDa subunit, CAF-I p150, chromatin assembly factor 1 subunit A, chromatin assembly factor 1, subunit A (p150), chromatin assembly factor I (150 kDa), Chromatin assembly factor I p150 subunit, hp150, MGC71229, P150 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CHAF1A (Q13111). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title