missing translation for 'onlineSavingsMsg'
Learn More

CHAF1A Rabbit anti-Human, Clone: 1T1O5, Novus Biologicals™

Product Code. 18322902
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322902 20 μg 20µL
18356215 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18322902 Supplier Bio-Techne Supplier No. NBP31669020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CHAF1A Monoclonal antibody specifically detects CHAF1A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHAF1A
Applications Western Blot
Classification Monoclonal
Clone 1T1O5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CAF, CAF1, CAF-1, CAF-1 subunit A, CAF1B, CAF1P150CAF-I 150 kDa subunit, CAF-I p150, chromatin assembly factor 1 subunit A, chromatin assembly factor 1, subunit A (p150), chromatin assembly factor I (150 kDa), Chromatin assembly factor I p150 subunit, hp150, MGC71229, P150
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CHAF1A (Q13111).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10036
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.