missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChABC Antibody (1E10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£183.00 - £387.00
Specifications
| Classification | Monoclonal |
|---|---|
| Purification Method | Protein G purified |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Molecular Weight of Antigen | 80 kDa |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18759723
|
Novus Biologicals
NBP1-96141SS |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18018324
|
Novus Biologicals
NBP1-96141 |
0.1 mL |
£387.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
| Monoclonal | |
| Protein G purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 80 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title