missing translation for 'onlineSavingsMsg'
Learn More

ChABC Antibody (1E10), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NBP1-96141

 View more versions of this product

Product Code. 18018324

  • £387.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ChABC
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Mouse
80 kDa
0.1 mL
Neurodegeneration, Neuroscience
Non-species specific
Purified
Western Blot, ELISA
10000000000
Unconjugated
Chondroitinase ABC
Chondroitinase ABC
Protein G purified
RUO
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only