missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CH 223191

Product Code. 18075892
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075892 50 mg 50mg
18004132 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075892 Supplier Tocris Bioscience™ Supplier No. 3858/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent aryl hydrocarbon receptor (AhR) antagonist

Potent aryl hydrocarbon receptor (AhR) antagonist (IC50 = 30 nM). Exhibits no AhR agonist-like activity (at concentrations up to 100 μM). Inhibits 2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD)-induced AhR-dependent transcription in vitro and reduces TCDD-induced toxicity in vivo. Attenuates Th17 differentiation of naive CD4 T cells and promotes expansion of hematopoietic stem cells in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301326-22-7
Molecular Formula C19H19N5O

Specifications

Quantity 50 mg
Formula Weight 333.39
Purity >98%
Molecular Weight (g/mol) 333.395
Chemical Name or Material CH 223191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.