CAS RN 301326-22-7
CAS RN 301326-22-7
IUPAC Name:
1-methyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}-1H-pyrazole-5-carboxamide
Synonyms:
2-methyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}pyrazole-3-carboxamide
Molecular Weight (g/mol):
333.4
Molecular Formula:
C19H19N5O
InChi Key:
LKTNEXPODAWWFM-UHFFFAOYSA-N
SMILES:
CN1N=CC=C1C(=O)NC1=C(C)C=C(C=C1)N=NC1=CC=CC=C1C
1
–
4
of
4
results
1
CH 223191, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ CH 223191
CAS: 301326-22-7 Molecular Formula: C19H19N5O
3
CH 223191, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
CH-223191, MedChemExpress
MedChemExpress CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR). CH-223191 inhibits TCDD-mediated nuclear translocation and DNA binding of AhR, and inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM.