missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CH 223191
Description
Potent aryl hydrocarbon receptor (AhR) antagonist (IC50 = 30 nM). Exhibits no AhR agonist-like activity (at concentrations up to 100 μM). Inhibits 2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD)-induced AhR-dependent transcription in vitro and reduces TCDD-induced toxicity in vivo. Attenuates Th17 differentiation of naive CD4 T cells and promotes expansion of hematopoietic stem cells in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 333.39 |
| Purity | >98% |
| Molecular Weight (g/mol) | 333.395 |
| Chemical Name or Material | CH 223191 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?