missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGRP Mouse anti-Human, Rat, Clone: 5, Novus Biologicals™
Mouse Monoclonal Antibody
£190.00 - £452.00
Specifications
| Antigen | CGRP |
|---|---|
| Clone | 5 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18023486
|
Novus Biologicals
NBP3-00518-0.025MG |
0.025 mg |
£190.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18019743
|
Novus Biologicals
NBP3-00518-0.1MG |
0.1 mg |
£452.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CGRP Monoclonal specifically detects CGRP in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CGRP | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
| Synthetic human alpha-CGRP | |
| Primary | |
| Store at 4°C in the dark. |
| 5 | |
| Western Blot, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 796 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title