missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGRP Mouse anti-Human, Rat, Clone: 5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-00518-0.1MG
This item is not returnable.
View return policy
Description
CGRP Monoclonal specifically detects CGRP in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CGRP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
| Synthetic human alpha-CGRP | |
| 0.1 mg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 5 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 796 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction