missing translation for 'onlineSavingsMsg'
Learn More

CGP 39551, Tocris Bioscience™

Product Code. 30001247
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001247

Brand: Tocris Bioscience 1409/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, selective and competitive NMDA antagonist

Potent, selective and competitive NMDA antagonist (Ki = 310 nM for inhibition of [3H]-CPP binding in rat brain). Centrally active upon oral administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127910-32-1
Molecular Formula C8H16NO5P
Molecular Weight (g/mol) 237.192
InChI Key OKDOWCKDTWNRCB-GQCTYLIASA-N
Synonym 2-amino-4-methyl-5-phosphono-3 pentenoic acid carboxyethyl ester, e-+/--2-amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, 1-ethyl ester, 3e, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, 1-ethyl ester, e-+-, 2e-4-amino-5-ethoxy-2-methyl-5-oxopent-2-en-1-ylphosphonic acid, cgp hplc, e-2-amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester, e-4-amino-5-ethoxy-2-methyl-5-oxopent-2-enyl phosphonic acid
PubChem CID 6372334
IUPAC Name [(E)-4-amino-5-ethoxy-2-methyl-5-oxopent-2-enyl]phosphonic acid
SMILES CCOC(=O)C(C=C(C)CP(=O)(O)O)N

Specifications

Quantity 10 mg
Formula Weight 237.19
Percent Purity >98%
Chemical Name or Material CGP 39551
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.