accessibility menu, dialog, popup

UserName

CAS RN 127910-32-1

H3COONH2CH3POHOHO(E)

CAS RN 127910-32-1

IUPAC Name: [(2E)-4-amino-5-ethoxy-2-methyl-5-oxopent-2-en-1-yl]phosphonic acid
Synonyms: (2E)-4-amino-5-ethoxy-2-methyl-5-oxopent-2-en-1-ylphosphonic acid
Molecular Weight (g/mol): 237.19
Molecular Formula: C8H16NO5P
InChi Key: OKDOWCKDTWNRCB-GQCTYLIANA-N
SMILES: CCOC(=O)C(N)\C=C(/C)CP(O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results