missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI-16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74084
This item is not returnable.
View return policy
Description
CGI-16 Polyclonal specifically detects CGI-16 in Human samples. It is validated for Western Blot.
Specifications
| CGI-16 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ACATE2, Acyl-CoA thioester hydrolase 9, acyl-CoA thioesterase 9acyl-Coenzyme A thioesterase 2, mitochondrial, acyl-coenzyme A thioesterase 9, mitochondrial, CGI-16, EC 3.1.2, EC 3.1.2.-, EC 3.1.2.15, mitochondrial Acyl-CoA Thioesterase, MT-ACT48 | |
| Rabbit | |
| 23 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Pig: 86%; Horse: 86%; Guinea pig: 86%; Rat: 79%; Bovine: 79%. | |
| Human, Rat, Bovine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96EA2 | |
| ACOT9 | |
| Synthetic peptides corresponding to the C terminal of ACOT9. Immunizing peptide sequence FLSSQVCFTQNNYIQVRVHSEVASLQEKQHTTTNVFHFTFMSEKEVPLVF. | |
| Affinity purified | |
| RUO | |
| 23597 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction