missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ CFSE
Click to view available options
Quantity:
5 x 500 μg
Unit Size:
Pack of 5
Description
- Molecular weight of 557.47g/mol
- After the acetate groups are cleaved, it has a peak excitation of 494nm and peak emission of 521nm
- Each vial may be reconstituted to stock concentration of 10 mM with 90μL of anhydrous DMSO
- Once reconstituted it should be used within 6 months and protected from light and stored at −20°C with dessicant; avoid freeze-thawing
- Unit Size: 5 x 500μg
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | CFSE |
| Emission | 494/521 |
| Format | 5 x 500 μg |
| Product Line | eBioscience |
| Quantity | 5 x 500 μg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction