missing translation for 'onlineSavingsMsg'
Learn More

Cesium dichromate, 99.8% (metals basis), Thermo Scientific™

Product Code. 11359387
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359387 100 g 100g
11369387 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359387 Supplier Thermo Scientific Alfa Aesar Supplier No. 040230.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13530-67-1
Molecular Formula Cr2Cs2O7
Molecular Weight (g/mol) 481.80
MDL Number MFCD00049436
InChI Key AQWIYQPYXQGBQU-UHFFFAOYSA-N
Synonym cesium dichromate, caesium dichromate, acmc-20alhd, cr2o7.2cs, dicaesium 1+ chromate, chromic acid h2cr2o7 ,cesium salt 1:2, dicesium oxido-oxido dioxo chromio oxy-dioxochromium
PubChem CID 3084158
IUPAC Name dicesium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [Cs+].[Cs+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Physical Form Coarse Powder
Quantity 100 g
Linear Formula Cs2Cr2O7
UN Number UN3087
Formula Weight 481.8
Percent Purity 99.8%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium dichromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.