CAS RN 13530-67-1
IUPAC Name:
dicaesium(1+) [(oxidodioxochromio)oxy]chromiumoylolate
Synonyms:
dicaesium(1+) chromate
Molecular Weight (g/mol):
481.8
Molecular Formula:
Cr2Cs2O7
InChi Key:
AQWIYQPYXQGBQU-UHFFFAOYSA-N
SMILES:
[Cs+].[Cs+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
1
–
1
of
1
results
1
Cesium dichromate, 99.8% (metals basis), Thermo Scientific™
CAS: 13530-67-1 Molecular Formula: Cr2Cs2O7 Molecular Weight (g/mol): 481.80 MDL Number: MFCD00049436 InChI Key: AQWIYQPYXQGBQU-UHFFFAOYSA-N Synonym: cesium dichromate,caesium dichromate,acmc-20alhd,cr2o7.2cs,dicaesium 1+ chromate,chromic acid h2cr2o7 ,cesium salt 1:2,dicesium oxido-oxido dioxo chromio oxy-dioxochromium PubChem CID: 3084158 IUPAC Name: dicesium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [Cs+].[Cs+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O