missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERCAM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CERCAM |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16866024
|
Proteintech
16411-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16856024
|
Proteintech
16411-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Has apparently no beta-galactosyltransferase activity.Specifications
| CERCAM | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| A3KGW5, Q5T4B2, Q5U309 | |
| 296616, 51148, 99151 | |
| CERCAM Fusion Protein Ag9352 | |
| Primary | |
| -20°C | |
| CERCAM |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CEECAM1, CERCAM, GLT25D3, KIAA1502 | |
| CERCAM | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title