missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERCAM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16411-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Has apparently no beta-galactosyltransferase activity.Specifications
| CERCAM | |
| Polyclonal | |
| Unconjugated | |
| CERCAM | |
| CEECAM1, CERCAM, GLT25D3, KIAA1502 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296616, 51148, 99151 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A3KGW5, Q5T4B2, Q5U309 | |
| CERCAM | |
| CERCAM Fusion Protein Ag9352 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction