Learn More
Eppendorf™ Centrifuge Services - IQ/OQ - GxP
Product Code. 17349371 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Multi-purpose centrifuge without refrigeration
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17349371 Multi-purpose centrifuge without refrigeration Each
1 options
Product Code. 17349371 Supplier Eppendorf™ Supplier No. 0082020302
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
GxP compliant IQ/OQ documentation support the complex GxP standards and regulations reducing your administrative workload, and provide you with qualified assurance that your instrument is installed and is operating in accordance with manufacturers' specifications.
  • Check of delivery status, and installation environment
  • Installation of instrument incl. configuration and functional check
  • Detailed IQ report
  • Test of alarm functions
  • Verification of instrument parameters
  • Detailed OQ report
  • Handling of deviation list/management
  • Signed approval by customer and service technician
TRUSTED_SUSTAINABILITY
Specifikationer
For Use With (Equipment) Multi-purpose centrifuge without refrigeration
Service Offering GxP Compliant IQ/OQ Documentation
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.