missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centaurin alpha 1 Antibody (3E2D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | Centaurin alpha 1 |
|---|---|
| Clone | 3E2D9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229671
|
Novus Biologicals
NBP2-61732 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694596
|
Novus Biologicals
NBP2-61732-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Centaurin alpha 1 Monoclonal specifically detects Centaurin alpha 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Centaurin alpha 1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 11033 | |
| Purified recombinant fragment of human ADAP1 (AA: 240-370) expressed in E. Coli. | |
| Primary | |
| 43.4 kDa |
| 3E2D9 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25 | |
| ADAP1 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title