missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centaurin alpha 1 Antibody (3E2D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61732
This item is not returnable.
View return policy
Description
Centaurin alpha 1 Monoclonal specifically detects Centaurin alpha 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Centaurin alpha 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25 | |
| Mouse | |
| 43.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 3E2D9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ADAP1 | |
| Purified recombinant fragment of human ADAP1 (AA: 240-370) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 11033 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction