missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPA Antibody (5A7-2E11), Novus Biologicals™
Mouse Monoclonal Antibody
£154.00
Specifications
| Antigen | CENPA |
|---|---|
| Clone | 5A7-2E11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
CENPA Monoclonal specifically detects CENPA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CENPA | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
| CENPA | |
| IgG1 | |
| Protein G purified | |
| Detects approx 18kDa. Recognizes Human CENP-A. |
| 5A7-2E11 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_001035891.1 | |
| 1058 | |
| Synthetic peptide corresponding to a portion of human CENP-A | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title