missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPA Antibody (5A7-2E11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12892-0.025mg
This item is not returnable.
View return policy
Description
CENPA Monoclonal specifically detects CENPA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CENPA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence | |
| Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1058 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 5A7-2E11 | |
| Unconjugated | |
| NP_001035891.1 | |
| CENPA | |
| Synthetic peptide corresponding to a portion of human CENP-A | |
| 0.025 mg | |
| Primary | |
| Detects approx 18kDa. Recognizes Human CENP-A. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction