missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CEM with CTA
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
Achieve precise microbial isolation with CEM Medium enriched with CTA, ideal for targeted cultivation of fastidious organisms.
Specifications
Specifications
| Color | Transparent |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Quantity | 10 Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction