Learn More
R&D Systems™ CellXVivo Human Treg Cell Differentiation Kit
Product Code. 18431420
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18431420 1 Kit 1 set
1 options
Product Code. 18431420
Brand: R&D Systems™ CDK006
For the differentiation of human regulatory T (Treg) cells from a preparation of naïve CD4+ T cells.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Includes Mouse Anti-Human CD3 Antibody
Human Treg Reagent 1
Human Treg Reagent 2
Human Treg Reagent 3
Human Treg Reagent 4
Reconstitution Buffer 1
Reconstitution Buffer 2
Wash Buffer (20X)
Quantity 1 Kit