missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellVue™ NIR815 Cell Labeling Kit
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
- The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
- CellVue™ NIR815 is optimally excited at 786nm and has a peak emission of 814nm
- CellVue™ NIR815 has been reported to be useful for short-term in vivo cell tracking studies using a Pearl™ Imager or Odyssey Infrared Imaging System (Li-COR Biosciences, Inc., Lincoln, NE)
Specifications
Specifications
| Quantity | 1 kit |
| Reagent Type | Cell Tracker Compounds, Cell Labeling Reagents |
| Product Type | Cell Labeling |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction