Learn More
Invitrogen™ CellVue™ NIR780 Cell Labeling Kit
Product Code. 15570847
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Product Code. 15570847
Brand: Invitrogen™ 88087516
Near-infrared fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • Midi CellVue™ Kits are supplied with two vials of dye stock (1 mM in ethanol) and six vials of Diluent C
  • CellVue™ NIR780 can be excited off the red laser line (633nm) and has a peak emission of 776nm that can be detected in filter sets designed for APC-eFluor™ 780.
  • Compatible with most multi-color applications, however, due to similar emission properties it cannot be used in combination with APC-eFluor™ 780 or APC-Alexa Fluor™ 750 reagents
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Label Type eFluor
Product Type Cell Labeling