missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellVue™ Lavender Cell Labeling Kit
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
- The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
- CellVue™ Lavender is optimally excited at 420nm and has a peak emission of 461nm
- CellVue™ Lavender is compatible with most multi-color flow cytometric applications; however, due to similar emission properties it cannot be used in combination with eFluor™ 450 or Pacific Blue™ reagents
Specifications
Specifications
| Quantity | 1 kit |
| Reagent Type | Cell Tracker Compounds, Cell Labeling Reagents |
| Product Type | Cell Labeling |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction