Learn More
Invitrogen™ CellVue™ Jade Cell Labeling Kit
Product Code. 15580847
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Product Code. 15580847
Brand: Invitrogen™ 88087616
Green fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ Jade is optimally excited at 478nm and has a peak emission of 508nm
  • Compatible with most multi-color applications; however, due to similar emission properties it cannot be used in combination with FITC or Alexa Fluor™ 488 reagents
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Product Type Cell Labeling