missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellVue Diluent C (For CellVue™ Cell Labeling Kits)
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Description
Diluent C is the Buffer required for the dilution of the dyes in the CellVue™ Cell Labeling Kits. Please refer to the specific labeling kit for instructions of use.
Reported Application
Flow Cytometric Analysis, Microscopy
Reported Application
Flow Cytometric Analysis, Microscopy
Specifications
Specifications
| Quantity | 10 mL |
| Validated Application | Cell Analysis, Flow Cytometry |
| Form | Kit |
| Reagent Type | Diluent |
| Product Type | Cell Labeling |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction