missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellVue™ Burgundy Cell Labeling Kit
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
- The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
- CellVue™ Burgundy is optimally excited at 683nm and has a peak emission of 707nm
- It can be detected equally in filter sets designed for Alexa Fluor™ 700 and APC-eFluor™ 780 or similar fluorochromes – using antibodies conjugated to these fluorochromes in combination with CellVue™ Burgundy is not recommended
- Burgundy can be used in combination with APC
Specifications
Specifications
| Quantity | 1 kit |
| Reagent Type | Cell Tracker Compounds, Cell Labeling Reagents |
| Label Type | eFluor |
| Product Type | Cell Labeling |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction