missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cellulose Membranes

Product Code. 11725315 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
Bubble Point:
3.1bar
4bar
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bubble Point unitSize
11725315 47 mm 3.1bar Pack of 100
11715315 25 mm 3.1bar Pack of 100
11705315 47 mm 4bar Pack of 100
11795305 25 mm 4bar Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11725315
Only null left
Add to basket
Add to basket

Suitable for Clarification and sterilizing filtration of biological fluids . Fisherbrand™ Cellulose Membranes filters are composed of pure cellulose acetate modified to offer researchers the lowest binding filters available.

  • Filtration sterile of aqueous solutions (nutrient media, sera, buffers)
  • Sterilizing filtration of proteins and enzymes
  • Provide high throughputs
  • Reduces filter changes when filtering proteinaceous solutions
  • Has Receptor binding studies
  • Low binding characteristics
  • Made of hydrophilic membranes
  • Uniform pore size structure allows consistent flow rates
  • Present strength and dimension stability (130psi) and resist to 135°C

Specifications

Bubble Point 3.1bar
Color White
Filter Type Microfilter
Material Cellulose Acetate
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 40mL/min./cm2
For Use With (Application) Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies
Gas Flow Rate 25L/min./cm2
Hydrophobicity Hydrophilic
Quantity 100 Pack
Type Plain Disc
Wettability Yes
Diameter (Metric) 47 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.