Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Product Code. 10022751 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inside:
15 mm
22 mm
31 mm
Height (Metric):
80 mm
100 mm
118 mm
180 mm
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Height (Metric) unitSize
10022751 31 mm 118 mm Pack of 25
10620303 22 mm 80 mm Pack of 25
10422032 31 mm 180 mm Pack of 25
10536101 15 mm 100 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10022751 Supplier Macherey-Nagel™ Supplier No. 645024
Only null left
Add to basket
Add to basket
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Quantity 25 Pk.
Diameter (Metric) Inside 31 mm
Material Cellulose
For Use With (Application) Analysis of soil contaminants, Analysis of soil contaminants
Height (Metric) 118 mm
Product Type Extraction Thimbles