missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellTrace™ Oregon Green™ 488 Carboxylic Acid Diacetate, Succinimidyl Ester (Carboxy-DFFDA, SE), cell permeant, mixed isomers
Description
Like CFDA SE, CellTrace™ Oregon Green™ 488 (carboxy-DFFDA SE) should be a useful tool for following proliferating cells. This Oregon Green™ 488 probe passively diffuses into cells, where it is colorless and nonfluorescent until its acetate groups are removed by intracellular esterases to yield a highly fluorescent, amine-reactive dye. Upon reaction with intracellular amines, the probe forms Oregon Green™ 488 conjugates that are well-retained by cells. Unlike fluorescein conjugates, Oregon Green™ 488 conjugates exhibit bright, photostable green fluorescence that is independent of pH at typical cellular pH values (pH-6-8).
Specifications
Specifications
| Content And Storage | Contains 20 vials of Oregon Green™ 488 carboxylic acid diacetate, SE (50 μg/vial). Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Carboxy-DFFDA SE |
| Dye Type | Oregon Green™ 488 |
| Form | Solid |
| Product Line | CellTrace, Oregon Green |
| Quantity | 20 x 50 μg |
| Reagent Type | Cell Tracker Compounds, Cell Labeling Reagents |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?