missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CellTrace™ Calcein Blue, AM - Special Packaging

Product Code. 11524277
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
1000µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11524277

Brand: Molecular Probes™ C34853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CellTrace™ calcein blue, AM - special packaging

CellTrace calcein blue AM is a proven short-term, blue-fluorescent tracer for cell labeling. The probe possesses acetoxymethyl (AM) esters that allow its passive diffusion across cell membranes. Upon cleavage of the AM esters by intracellular esterases, the molecule that remains (absorption/emission maxima ∼360/449 nm) is relatively polar and is retained by cells for several hours.

View a selection guide for all Nonfixable Viability Dyes for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 20 vials of calcein blue (50 μg per vial). Store in freezer (-5 to -30°C).
Excitation Wavelength Range 323⁄439
Dye Type calcein blue
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Quantity 20 x 50 μg
Detection Method Fluorescence
Emission UV
Form Solid
Product Line CellTrace
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Label Type Other Labels or Dyes
Product Type Viability Indicator
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.