Learn More
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks
GREENER_CHOICE
Product Code. 10116520
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
650 mL
Closure Type:
Screw
White Filter Screw Cap
Unit Size:
Pack of 120
Pack of 200
Pack of 40
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Type unitSize
10116520 250 mL Screw Pack of 120
10716172 250 mL White Filter Screw Cap Pack of 120
10507071 650 mL White Filter Screw Cap Pack of 40
10722541 50 mL Screw Pack of 200
10668821 50 mL White Filter Screw Cap Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10116520 Supplier Greiner Bio-One Supplier No. 658190
Add to basket
Add to basket
Designed to culture cell suspended in a growth medium. Greiner Bio-One™ CELLSTAR Suspension Cell Culture Flasks are manufactured from polystyrene for optimum visibility and strength.
  • Hydrophobic surface, ideal for suspension cultures and hybridoma
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Graduation on the side
  • Canted neck
  • Easily distinguished by their white screw caps
  • White standard screw cap with ventilation position
  • TC surface treatment: -/suspension
  • Sterile and user friendly packaging
TRUSTED_SUSTAINABILITY
Specifications
Closure Color White
Capacity (Metric) 250 mL
Material Polystyrene
No. per Case 120 Pack
No. per Pack 120
Graduated Yes
Closure Type Screw