missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ SIE-bla ME-180 Cell Line

Product Code. 10584424
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10584424

Brand: Thermo Scientific™ K1648

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The CellSensor&tarde; SIE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Sis-Inducible Element (SIE) stably integrated into ME-180 cells.

The CellSensor&tarde; SIE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Sis-Inducible Element (SIE) stably integrated into ME-180 cells. To construct this cell line, the SIE-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Interleukin-6 (IL-6). This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations of IL-6. The CellSensor™ SIE-bla ME-180 cell line is responsive to IL-6, OSM and IFNγ and can be used to probe the JAK/STAT signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Signaling Pathway
Product Line CellSensor
Quantity 1 Vial
Target Entry STAT1, STAT3, JAK/STAT (IFN gamma, IL-6/OSM) pathway, JAK2, JAK1
Type Dividing Cells
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.