Learn More
Thermo Scientific™ CellSensor™ SBE-bla A375 Cell Line
Description
Bone morphogenetic proteins (BMPs), members of the transforming growth factor beta (TGF-β) superfamily, play important roles in the development of the heart, central nervous system and cartilage. Disruption of BMP signaling affects the body plan of the developing embryo. BMPs propagate their signal by activating Activin receptor-like kinase (ALK), which in turn mediates the phosphorylation of R-Smads. Phosphorylated R-Smad associates with Smad4 and then translocates to the nucleus regulating gene expression. A375 cells are human malignant melanoma cells containing endogenous Smad signaling pathway. SBE-bla A375 cell line was engineered to express beta-lactamase under the control of Smad binding element. This cell line has been validated for Z' and EC50 under optimized conditions using BMP-4. This cell line has also responded to Nodal and was tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, substrate loading time. Additional information using various small molecule inhibitors and StealthTM RNAi is also provided.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ SBE-bla A375 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | Cell Line: A375 |
| Detection Method | Fluorescence |
| For Use With (Application) | Signaling Pathway |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | ALK5, ALK7, TGF beta/BMP pathway |
| Type | Dividing Cells |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.