Learn More
Thermo Scientific™ CellSensor™ NFkB-bla ME-180 Cell Line
Description
The CellSensor™ NFκB-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Nuclear Factor Kappa B (NFκB) response element stably integrated into ME-180 cells. The cell line was created through fluorescence activated cell sorting (FACS) of cells responsive to stimulation of the NFκB pathway with tumor necrosis factor alpha (TNFα). The cell line was validated for DMSO tolerance, incubation time with stimulant, and substrate loading conditions. The CellSensor™ NFκB-bla ME-180 cell line responds to agonist treatment as expected from literature and can be adapted for high-throughput screening for agonists or antagonists of the NFκB pathway with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ NFkB-bla ME-180 Cell Line, 1Vial, ∽ 9,600,000 cells/vial at 9,600,000 cells/mL Quantity of Cells, NFkB-bla ME-180 Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays |
| Includes | beta-lactamase reporter gene under control of the nuclear factor Kappa B (NFkB) response element stably integrated into ME-180 cells |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | NFkB-bla ME-180 |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.