missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CellLight™ Nucleus-CFP, BacMam 2.0

Product Code. 11602099
Click to view available options
Quantity:
1 vial(s)
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 11602099

Brand: Molecular Probes™ C10616

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CellLight™ Nucleus-CFP, BacMam 2.0

CellLight™ Nucleus-CFP, BacMam 2.0, provides an easy way to label nuclei with cyan fluorescent protein (CFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight™ fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses CFP fused to the SV40 nuclear localization sequence. You can observe nucleus-CFP behavior in live cells without the cellular toxicity associated with intercalators and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight™ constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight™ Technology is:
  • Fast and convenient: simply add CellLight™ reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Less toxic: CellLight™ reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

    BacMam Technology

    CellLight™ Nucleus-CFP, BacMam 2.0, is a fusion construct of SV40 nuclear localization sequence and CFP, providing accurate and specific targeting to cellular nucleus-CFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight™ reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color Blue
    Content And Storage Store at 2°C to 6°C, protected from light. Do Not Freeze.
    Dye Type CFP (Cyan Fluorescent Protein)
    For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
    Quantity 1 vial(s)
    Detection Method Fluorescence
    Emission Visible
    Form Liquid
    Product Line CellLight, Molecular Probes
    Shipping Condition Wet Ice
    Technique Fluorescence Intensity
    Label Type Fluorescent Dye
    Product Type Nucleus Label
    SubCellular Localization Nucleus
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.