Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
GREENER_CHOICEBIOPHARMA
Product Code. p-7221304 Shop All Corning Products
Click to view available options
Tray Layers:
10-Layer
100-Layer
25-Layer
Unit Size:
Each
Pack of 2
Product Code. 17299099
Brand: Corning™ 3231
Only null left
Add to basket
Add to basket
Provides a fast, simple, and compact method for the mass culture of attachment-dependent cells.
  • Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
  • The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
  • The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
TRUSTED_SUSTAINABILITY
Specifications
Includes Growth Surface and Aseptic Connectors
No. per Case 2/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Culture Area 8500 cm²
Tray Layers 10-Layer