missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 670
A red fluorescent dye that can be used to monitor individual cell divisions for up to 6 generations.
Brand: Invitrogen™ 65-0840-90
Code : NEW
Additional Details : Weight : 0.09500kg
Description
- A red fluorescent dye that can be used to monitor individual cell divisions
- Dye binds to any cellular protein containing primary amines, and as cells divide, it is distributed equally between daughter cells that can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 6 generations
- Cell Proliferation Dye eFluor™ 670 can also be used for long term tracking of labeled cells
- Analysis using two-parameter plots may provide better resolution of each generation, especially between undivided cells and the first generation
- Cells labeled with Cell Proliferation Dye eFluor™ 670 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
- Cell Proliferation Dye eFluor™ 670 has a peak excitation of 647 nm and can be excited by the red (633 nm) laser line
- It has a peak emission of 670 nm and can be detected with a 660/20 band pass filter (equivanet to APC, Alexa Fluor™ 647, or eFluor™ 660), making it compatible with applications that utilize GFP
Additional Details
- Cell Proliferation Dye eFluor™ 670 has a molecular weight of 792.6
- Each vial may be reconstituted to a stock concentration of 5 mM with 126 μL of anhydrous DMSO
- Once reconstituted it should be protected from light and stored at -20°C with dessicant
- It is recommended to use the reconstituted dye within 6 months and to avoid freeze-thawing
Specifications
Red | |
Proliferation Assay | |
Dye | |
Lyophilized powder | |
4 x 500 μg |
Fluorescent | |
Fluorescence Microscope, Flow Cytometer | |
630/670 | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.