missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 670

Product Code. 15304860
Change view
Click to view available options
Quantity:
4 x 500 μg
500 μg
Unit Size:
500µg
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15304860 500 μg 500µg
15500597 4 x 500 μg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15304860 Supplier Invitrogen™ Supplier No. 65084085
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

A red fluorescent dye that can be used to monitor individual cell divisions for up to 6 generations.

  • A red fluorescent dye that can be used to monitor individual cell divisions
  • Dye binds to any cellular protein containing primary amines, and as cells divide, it is distributed equally between daughter cells that can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 6 generations
  • Cell Proliferation Dye eFluor™ 670 can also be used for long term tracking of labeled cells
  • Analysis using two-parameter plots may provide better resolution of each generation, especially between undivided cells and the first generation
  • Cells labeled with Cell Proliferation Dye eFluor™ 670 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
  • Cell Proliferation Dye eFluor™ 670 has a peak excitation of 647 nm and can be excited by the red (633 nm) laser line
  • It has a peak emission of 670 nm and can be detected with a 660/20 band pass filter (equivanet to APC, Alexa Fluor™ 647, or eFluor™ 660), making it compatible with applications that utilize GFP

Additional Details

  • Cell Proliferation Dye eFluor™ 670 has a molecular weight of 792.6
  • Each vial may be reconstituted to a stock concentration of 5 mM with 126 μL of anhydrous DMSO
  • Once reconstituted it should be protected from light and stored at -20°C with dessicant
  • It is recommended to use the reconstituted dye within 6 months and to avoid freeze-thawing
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Proliferation Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Emission 630/670
Form Lyophilized powder
Product Line eBioscience
Quantity 500 μg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.