missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 450
Click to view available options
Quantity:
4 x 500 μg
500 μg
Unit Size:
500µg
Pack of 4
Description
- A violet fluorescent dye that can be used to monitor individual cell divisions
- Dye binds to any cellular proteins containing primary amines, and as cells divide, the dye is distributed equally between daughter cells
- They can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 7 generations
- Cells labeled with Cell Proliferation Dye eFluor™ 450 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
- Cell Proliferation Dye eFluor™ 450 has a peak excitation of 409 nm and can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™), making it compatible with applications that utilize GFP
Additional Details
- Each vial may be reconstituted to a stock concentration of 10 mM with 165 μL of anhydrous DMSO
- Once reconstituted it should be used within 6 months and protected from light and stored at -20°C with dessicant
- Avoid freeze-thawing
Specifications
Specifications
| Color | Violet |
| Detection Method | Fluorescence |
| For Use With (Application) | Proliferation Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Dye |
| Emission | 405/450 |
| Form | Lyophilized powder |
| Product Line | eBioscience |
| Quantity | 4 x 500 μg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction