Learn More
Agilent BioTek Cell Count & Viability Starter Kit
Product Code. 17140549 Shop All Agilent BioTek Products
Click to view available options
:
Cell Count & Viability Starter Kit
Product Code. 17140549
Brand: Agilent BioTek 1980001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cell Count & Viability Starter Kit
  • Includes:
    • Cell Count & Viability App: Pre-defined protocols automatically calculate count and percent live/dead
    • Box of 50 disposable counting slides (2 samples/slide)
    • 1.5 mL vial of 0.4% trypan blue for staining dead cells
    • 4-position counting slide holder
    • Instructions for use
    • 8 samples total
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cell Count and Viability Starter Kit
For Use With (Application) Cell Count