Learn More
Agilent BioTek Cell Count & Viability App software
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Agilent BioTek Microplate Washers and Readers
Unit Size:
Each
Description
- Simplified interface enabling rapid cell counts and analysis for up to 8 samples simultaneously
- Compatible with Lionheart FX and Cytation 5
- Specifically designed for automated mammalian cell counting and analysis
Specifications
Specifications
| For Use With (Equipment) | Agilent BioTek Microplate Washers and Readers |
| Type | Software |