missing translation for 'onlineSavingsMsg'
Learn More

Cefazedone, TRC

Product Code. 30363757
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363757 10 mg -
30357886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363757 Supplier Toronto Research Chemicals Supplier No. TRCC24150010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56187-47-4
Molecular Formula C18H15Cl2N5O5S3
Molecular Weight (g/mol) 548.44
Synonym (6R,7R)-7-[[2-(3,5-Dichloro-4-oxo-1(4H)-pyridinyl)acetyl]amino]-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R-trans)-Cefazedone, Refosporen
IUPAC Name (6R,7R)-7-[[2-(3,5-dichloro-4-oxopyridin-1-yl)acetyl]amino]-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)CN4C=C(Cl)C(=O)C(=C4)Cl)C3=O)C(=O)O)s1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H15Cl2N5O5S3/c1-7-22-23-18(33-7)32-6-8-5-31-16-12(15(28)25(16)13(8)17(29)30)21-11(26)4-24-2-9(19)14(27)10(20)3-24/h2-3,12,16H,4-6H2,1H3,(H,21,26)(H,29,30)/t12-,16-/m1/s1
Formula Weight 546.9612
Chemical Name or Material Cefazedone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.