CAS RN 56187-47-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefazedone, TRC
CAS: 56187-47-4 Molecular Formula: C18H15Cl2N5O5S3 Molecular Weight (g/mol): 548.44 Synonym: (6R,7R)-7-[[2-(3,5-Dichloro-4-oxo-1(4H)-pyridinyl)acetyl]amino]-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid,(6R-trans)-Cefazedone,Refosporen IUPAC Name: (6R,7R)-7-[[2-(3,5-dichloro-4-oxopyridin-1-yl)acetyl]amino]-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)CN4C=C(Cl)C(=O)C(=C4)Cl)C3=O)C(=O)O)s1